missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNA14 Polyclonal Antibody, Invitrogen™
Description
This gene encodes a member of the guanine nucleotide-binding, or G protein family. G proteins are heterotrimers consisting of alpha, beta and gamma subunits. The encoded protein is a member of the alpha family of G proteins, more specifically the alpha q subfamily of G proteins. The encoded protein may play a role in pertussis-toxin resistant activation of phospholipase C-beta and its downstream effectors.[provided by RefSeq, Feb 2009].
Specifications
Specifications
| Antigen | GNA14 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | GNA14 |
| Gene Accession No. | O95837, P30677 |
| Gene Alias | G-protein subunit alpha-14, g alpha-14, guanine nucleotide-binding protein 14, guanine nucleotide-binding protein subunit alpha-14 |
| Gene Symbols | GNA14 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?