Learn More
Multiroir Controlec™ GM Sectioned Drawer Inserts
Product Code. 11774571
Change view
Click to view available options
Array:
3 x 4
4 x 4
Width (Metric):
55 mm (compartment)
79 mm (compartment)
Length (Metric):
77 mm (compartment)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Array Width (Metric) Length (Metric) unitSize
11774571 3 x 4 79 mm (compartment) 77 mm (compartment) Each
11784571 4 x 4 55 mm (compartment) 77 mm (compartment) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11774571 Supplier Multiroir Controlec™ Supplier No. 32154
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Fits securely in the tray of a GM Drawer Cabinet. Multiroir Controlec™ GM Sectioned Drawer Inserts are thermoformed honeycomb grid pieces that customize the plastic drawer with the size and number of compartments needed for sectioned storage.
GM Drawer Cabinets are a block of six colored tray-drawers to suit a variety of sorted storage needs.
  • Solid polypropylene construction
  • Color coded trays / drawers
  • Transportable and stackable
  • Drawers can be equipped with a variety of honeycomb grids
  • Drawer/Insert Size: 340 x 250 x 40mm
Specifications
Material Plastic
Product Type Sectioned Drawer Insert
For Use With (Application) Sectioned storage
Holds 12 Compartments
Height (Metric) 40 mm
Color White
Width (Metric) 79 mm (compartment)
Length (Metric) 77 mm (compartment)
Array 3 x 4