missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GlyT2/SLC6A5 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33239-100ul
This item is not returnable.
View return policy
Description
GlyT2/SLC6A5 Monoclonal antibody specifically detects GlyT2/SLC6A5 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| GlyT2/SLC6A5 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| GlyT2, GLYT-2, GLYT2GlyT-2, NET1, sodium- and chloride-dependent glycine transporter 2, solute carrier family 6 (neurotransmitter transporter, glycine), member 5, Solute carrier family 6 member 5 | |
| A synthetic peptide corresponding to a sequence within amino acids 250-350 of human GlyT2/SLC6A5 (Q9Y345).,, Sequence:, SLGQFASQGPVSVWKAIPALQGCGIAMLIISVLIAIYYNVIICYTLFYLFASFVSVLPWGSCNNPWNTPECKDKTKLLLDSCVISDHPKIQIKNSTFCMTA | |
| 100 μL | |
| Neuroscience | |
| 9152 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction