missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GlyRS-IN-1, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
GlyRS-IN-1 is a glycyl-tRNA synthase (GlyRS) inhibitor extracted from patent WO 2017066459 A1. GlyRS-IN-1 can also inhibit the growth of bacteria.
Specifications
Specifications
| Chemical Name or Material | GlyRS-IN-1 |
| CAS | 112921-11-6 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C12H17N7O7S |
| Quantity | 5 mg |
| Solubility Information | DMSO : 100 mg/mL (247.91 mM; Need ultrasonic) |
| SMILES | O[C@@H]1[C@@H](COS(=O)(NC(CN)=O)=O)O[C@@H](N2C(N=CN=C3N)=C3N=C2)[C@@H]1O |
| Molecular Weight (g/mol) | 403.37 |
| Formula Weight | 403.37 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction