Learn More
Glyoxal dimethyl acetal, 60% aq. soln.
Product Code. p-7047275
Click to view available options
Quantity:
100 g
500 g
2000 g
Unit Size:
100g
2kg
500g
Product Code. 11315015
Brand: Thermo Scientific Alfa Aesar L18490.22
Request Bulk or Custom Format
CAS: 51673-84-8 | C4H8O3 | 104.105 g/mol
Glyoxal dimethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glyoxal dimethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuffs.
Solubility
Fully miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51673-84-8
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.105
MDL Number MFCD00134410
InChI Key OGFKTAMJLKHRAZ-UHFFFAOYSA-N
Synonym acetaldehyde, dimethoxy, dimethoxyacetaldehyde, dimethoxy acetaldehyde, glyoxal dimethylacetal, glyoxal dimethyl acetal, 60 wt. % in h2o, acetaldehyde, 2,2-dimethoxy, 2,2-dimethoxyethanal, glyoxaldimethylacetal, dimethoxyethanal
PubChem CID 162650
IUPAC Name 2,2-dimethoxyacetaldehyde
SMILES COC(C=O)OC
Specifications
Density 1.15
Boiling Point 100°C to 102°C
Flash Point 102°C (216°F)
Refractive Index 1.414
Quantity 100 g
Beilstein 1850744
Solubility Information Fully miscible in water.
Formula Weight 104.11
Concentration or Composition (by Analyte or Components) 60% aq. soln.
Chemical Name or Material Glyoxal dimethyl acetal