missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GLYCTK Polyclonal Antibody
GREENER_CHOICE

Product Code. 17742544
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17742544 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17742544 Supplier Invitrogen™ Supplier No. PA5121146

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse liver, Mouse kidney, Rat liver, Rat kidney. The target is usually found in the following locations: Cytoplasm, Cytoplasm, Mitochondrion. Immunogen sequence: VKTVLSRADS DPHGPHTCGH VLNVIIGSNV LALAEAQRQA EALGYQAVVL SAAMQGDVKS MAQFYGLLAH VARTRLTPSM AGASVEEDAQ LHELAAELQI PDLQLEEALE TMAWGRGPVC LLAGGEPTVQ LQGSGRGGRN QELALRVGAE LRRWPLGPID VLFLSGGTDG QDGPTEAAGA WVTPELASQA AAEGLDIATF LAHNDSHTFF CCLQGGAHLL HTGMTGTNVM DTHLLFLRPR.

This locus encodes a member of the glycerate kinase type-2 family. The encoded enzyme catalyzes the phosphorylation of (R)-glycerate and may be involved in serine degradation and fructose metabolism. Decreased activity of the encoded enzyme may be associated with the disease D-glyceric aciduria. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLYCTK
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GLYCTK
Gene Accession No. Q0VGK3, Q8IVS8, Q8QZY2
Gene Alias 6230410P16Rik; C430006L12; glycerate kinase; GLYCTK; HBeAg binding protein 4; HBeAg-binding protein 2; HBeAg-binding protein 4; HBeAgBP4A; HBEBP2; HBEBP4; LP5910
Gene Symbols GLYCTK
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 284-523 of human GLYCTK (NP_660305.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132158, 235582, 684314
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.