missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycine, sodium salt, 99%

Product Code. p-139060 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10318200

Brand: Thermo Scientific Chemicals 204591000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6000-44-8 | C2H4NNaO2 | 97.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6000-44-8
Molecular Formula C2H4NNaO2
Molecular Weight (g/mol) 97.05
MDL Number MFCD00150683,MFCD00062543
InChI Key WUWHFEHKUQVYLF-UHFFFAOYSA-M
Synonym sodium glycinate, sodium 2-aminoacetate, glycine sodium salt, glycine, monosodium salt, sodium glycine, sodium aminoacetate, sodium glycocollate, sodum glycinate, glycine, sodium salt, unii-f6b979bv8z
PubChem CID 4684308
SMILES [Na+].NCC([O-])=O

Specifications

Melting Point 197.0°C to 201.0°C
pH 11.1 to 12.0 (10% aq. soln.)
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula H2NCH2CO2Na
Transmittance 94.0 % min. (c=5, H2O, 400nm) 1 cm cell
Solubility Information Solubility in water: >500g/L (20°C). Other solubilities: solubility in methanol: 60g/L (20°C)
Water 1% Max. (K.F.)
IUPAC Name sodium;2-aminoacetate
Quantity 100 g
Formula Weight 97.05
Percent Purity 99%
Physical Form Moist Beads
Chemical Name or Material Glycine, sodium salt, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.