Learn More
Glycine, Molecular biology reagent, ≥99%, MP Biomedicals™
Product Code. p-4679927 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
500g
5kg
Product Code. 11426551
Brand: MP Biomedicals 0219482580_CLEAN
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Protein Electrophoresis & Blotting
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O
Specifications
Assay Percent Range ≥99%
Density 1.16g/mL (Lit.)
Quantity 100 g
Formula Weight 75.07
Grade Molecular Biology
Physical Form Powder
Chemical Name or Material Glycine