missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl Palmitate, TRC

Product Code. 17969064
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17969064 10 mg 10mg
17987604 100 mg 100mg
17928494 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17969064 Supplier Toronto Research Chemicals Supplier No. TRCG61595010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7501-44-2
Molecular Formula C19 H36 O3
Molecular Weight (g/mol) 312.49
Synonym Glycidyl Palmitate, Hexadecanoic acid, 2-oxiranylmethyl ester, Hexadecanoic acid, oxiranylmethyl ester (9CI), Palmitic acid, 2,3-epoxypropyl ester (8CI), (±)-Glycidyl palmitate, Glycidyl hexadecanoate, Glycidyl palmitate, NSC 406558, Palmitic acid glycidyl ester
IUPAC Name oxiran-2-ylmethyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC1CO1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H36O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(20)22-17-18-16-21-18/h18H,2-17H2,1H3
Formula Weight 312.2664
Percent Purity >95%
Chemical Name or Material Glycidyl Palmitate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.