missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl Palmitate (1.0 mg/mL in Acetonitrile), TRC

Product Code. 30464789
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464789 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464789 Supplier Toronto Research Chemicals Supplier No. TRCKIT979510X1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7501-44-2
Molecular Formula C19 H36 O3
Quantity 10 x 1 mL
Synonym Glycidyl Palmitate, Hexadecanoic acid, 2-oxiranylmethyl ester, Hexadecanoic acid, oxiranylmethyl ester (9CI), Palmitic acid, 2,3-epoxypropyl ester (8CI), (±)-Glycidyl palmitate, Glycidyl hexadecanoate, Glycidyl palmitate, NSC 406558, Palmitic acid glycidyl ester
InChI Formula InChI=1S/C19H36O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(20)22-17-18-16-21-18/h18H,2-17H2,1H3
SMILES CCCCCCCCCCCCCCCC(=O)OCC1CO1
IUPAC Name oxiran-2-ylmethyl hexadecanoate
Molecular Weight (g/mol) 312.49
Formula Weight 312.2664
Concentration or Composition (by Analyte or Components) Glycidyl Palmitate 1 mg/mL
Chemical Name or Material Glycidyl Palmitate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.