Learn More
Glycerol-3-phosphate oxidase, MP Biomedicals™
Product Code. 11409671 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 U
50 U
500 U
Unit Size:
100 units
50 units
500 units
Product Code. 11409671
Brand: MP Biomedicals 0215724090
Chemical Identifiers
CAS 9046-28-0
Synonym sn-Glycerol-3-phosphate, oxygen 2-oxidoreductase
Specifications
Color Yellow
Quantity 500 U
Physical Form Lyophilized Powder
Chemical Name or Material Glycerol-3-phosphate oxidase