missing translation for 'onlineSavingsMsg'
Learn More

Glycerol 1-Oleyl Ether, TRC

Product Code. 30136501
Change view
Click to view available options
Quantity:
10 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136501 100 mg 100mg
30134115 200 mg 200mg
17909844 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136501 Supplier Toronto Research Chemicals Supplier No. TRCG598710100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-31-7
Molecular Formula C21H42O3
Molecular Weight (g/mol) 342.56
Synonym 3-[9-Octadecen-1-yloxy]-1,2-propanediol, 3-(9-Octadecenyloxy)-1,2-propanediol, 3-[9-Octadecenyloxy]-1,2-propanediol, NSC 29230, NSC 36089, Selachyl alcohol, alpha-Glycerol monooleyl ether, alpha-Monooleyl glyceryl ether;
IUPAC Name 3-[(Z)-octadec-9-enoxy]propane-1,2-diol
SMILES CCCCCCCC\C=C/CCCCCCCCOCC(O)CO

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H42O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-24-20-21(23)19-22/h9-10,21-23H,2-8,11-20H2,1H3/b10-9-
Formula Weight 342.3134
Chemical Name or Material Glycerol 1-Oleyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.