Learn More
Glutathione Peroxidase 4/GPX4 Antibody (9D4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26545-100ul
 View more versions of this product
Product Code. 30043082
  • 533.00 EUR / 100µL
Description
Glutathione Peroxidase 4/GPX4 Monoclonal antibody specifically detects Glutathione Peroxidase 4/GPX4 in Human samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Glutathione Peroxidase 4/GPX4
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry
9D4
ELISA, Immunohistochemistry 1:50-1:200
EC 1.11.1, EC 1.11.1.12, Glutathione peroxidase 4, glutathione peroxidase 4 (phospholipid hydroperoxidase), GPx-4, GSHPx-4, MCSP, PHGPxsnGPx, phospholipid hydroperoxidase, phospholipid hydroperoxide glutathione peroxidase, mitochondrial, snPHGPx, sperm nucleus glutathione peroxidase
A synthesized peptide derived from Human Glutathione Peroxidase 4/GPX4 [UniProt P36969]
100 μL
Cancer, Endocrinology, Signal Transduction
2879
Store at -20 to -70C. Avoid freeze-thaw cycles.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions