Learn More
Glucosamine, MedChemExpress
Product Code. 16478626
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478626 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16478626 Supplier Medchem Express Supplier No. HYB1125
MedChemExpress Glucosamine (D-Glucosamine) is an amino sugar and a prominent precursor in the biochemical synthesis of glycosylated proteins and lipids, is used as a dietary supplement. Glucosamine also is a natural constituent of glycosaminoglycans in the cartilage matrix and synovial fluid, which when administered exogenously, exerts pharmacological effects on osteoarthritic cartilage and chondrocytes.
Glucosamine (D-Glucosamine) is an amino sugar and a prominent precursor in the biochemical synthesis of glycosylated proteins and lipids, is used as a dietary supplement. Glucosamine also is a natural constituent of glycosaminoglycans in the cartilage matrix and synovial fluid, which when administered exogenously, exerts pharmacological effects on osteoarthritic cartilage and chondrocytes.
Specifications
Chemical Name or Material Glucosamine
CAS 3416-24-8
Color White
Purity Grade Notes Research
Molecular Formula C6H13NO5
Quantity 100 mg
Synonym D-Glucosamine Chitosamine
Solubility Information H2O : 100 mg/mL (558.13 mM; Need ultrasonic) ∣DMSO : 3.33 mg/mL (18.59 mM; Need ultrasonic)
SMILES O=C[C@H](N)[C@H]([C@@H]([C@@H](CO)O)O)O
Molecular Weight (g/mol) 179.17
Formula Weight 179.17
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation