Learn More
Glucoraphanin, TRC
Specifications
Specifications
| CAS | 21414-41-5 |
| Molecular Formula | C12 H23 N O10 S3 |
| Quantity | 1 mg |
| Synonym | β-D-Glucopyranose, 1-thio-, 1-[5-(methylsulfinyl)-N-(sulfooxy)pentanimidate], Glucopyranose, 1-thio-, 1-[5-(methylsulfinyl)valerohydroximate] NO-(hydrogen sulfate), β-D- (8CI), 4-Methylsulfinylbutyl glucosinolate, Glucorafanin, Glucoraphanin, Sulforaphane glucosinolate |
| InChI Formula | InChI=1S/C12H23NO10S3/c1-25(18)5-3-2-4-8(13-23-26(19,20)21)24-12-11(17)10(16)9(15)7(6-14)22-12/h7,9-12,14-17H,2-6H2,1H3,(H,19,20,21)/t7-,9-,10+,11-,12+,25?/m1/s1 |
| SMILES | CS(=O)CCCCC(=NOS(=O)(=O)O)S[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O |
| IUPAC Name | [(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] 5-methylsulfinyl-N-sulfooxypentanimidothioate |
| Molecular Weight (g/mol) | 437.51 |
| Formula Weight | 437.0484 |
| Chemical Name or Material | Glucoraphanin |