Learn More
Glibornuride, TRC
Product Code. 17929744
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17929744 2.5 mg 2.5mg
17937624 5 mg 5mg
17967994 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17929744 Supplier Toronto Research Chemicals Supplier No. TRCG4097002.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 26944-48-9
Molecular Formula C18 H26 N2 O4 S
Molecular Weight (g/mol) 366.48
Synonym Benzenesulfonamide, N-[[[(1S,2S,3R,4R)-3-hydroxy-4,7,7-trimethylbicyclo[2.2.1]hept-2-yl]amino]carbonyl]-4-methyl-, Benzenesulfonamide, N-[[(3-hydroxy-4,7,7-trimethylbicyclo[2.2.1]hept-2-yl)amino]carbonyl]-4-methyl-, [1S-(endo,endo)]-, Urea, 1-(2-hydroxy-3-bornyl)-3-(p-tolylsulfonyl)-, (1R,2R,3S,4S)- (8CI), Glibornuride, Gluboride, Glutril, Ro 6-4563, Ro 6-4563/8, endo, endo-1-[(1R)-(2-Hydroxy-3-bornyl)]-3-(p-tolylsulfonyl)urea, endo,endo-1-(2-Hydroxy-3-bornyl)-3-(p-tolylsulfonyl)urea
IUPAC Name 1-[(1S,2S,3R,4R)-3-hydroxy-4,7,7-trimethyl-2-bicyclo[2.2.1]heptanyl]-3-(4-methylphenyl)sulfonylurea
SMILES Cc1ccc(cc1)S(=O)(=O)NC(=O)N[C@@H]2[C@H](O)[C@]3(C)CC[C@H]2C3(C)C
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C18H26N2O4S/c1-11-5-7-12(8-6-11)25(23,24)20-16(22)19-14-13-9-10-18(4,15(14)21)17(13,2)3/h5-8,13-15,21H,9-10H2,1-4H3,(H2,19,20,22)/t13-,14+,15+,18+/m1/s1
Formula Weight 366.1613
Percent Purity >95%
Chemical Name or Material Glibornuride
Grade HPLC