missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526838
This item is not returnable.
View return policy
Description
This antibody is predicted to react with chicken, hamster and rat based on sequence homology.
GLG1 is a protein coding gene. Binds fibroblast growth factor and E-selectin (cell-adhesion lectin on endothelial cells mediating the binding of neutrophils).
Specifications
| GLG1 | |
| Polyclonal | |
| Unconjugated | |
| GLG1 | |
| AI593353; AW537898; CFR; CFR1; CFR-1; Cysteine-rich fibroblast growth factor receptor; E-selectin ligand 1; ESL1; ESL-1; Glg1; Golgi apparatus protein 1; golgi glycoprotein 1; golgi sialoglycoprotein MG-160; MG160; MG-160; selectin, endothelial cell, ligand; Selel | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 20340, 2734 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q61543, Q92896 | |
| GLG1 | |
| KLH conjugated synthetic peptide between 1152-1179 amino acids from the C-terminal region of human GLG1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction