Learn More
Glecaprevir, TRC
Specifications
Specifications
| Format | Neat |
| Molecular Formula | C38H46F4N6O9S |
| Molecular Weight (g/mol) | 838.86, 838.87 |
| InChI Key | MLSQGNCUYAMAHD-ZSXAAYSXSA-N |
| SMILES | [H][C@@]12C[C@H](N(C1)C(=O)[C@@]([H])(NC(=O)O[C@]1([H])CCC[C@@]1([H])OC\C=C/C(F)(F)C1=NC3=CC=CC=C3N=C1O2)C(C)(C)C)C(=O)N[C@@]1(C[C@H]1C(F)F)C(=O)NS(=O)(=O)C1(C)CC1 |
| IUPAC Name | (1R,14Z,18R,22R,26S,29S)-26-tert-butyl-N-[(1R,2R)-2-(difluoromethyl)-1-{[(1-methylcyclopropyl)sulfonyl]carbamoyl}cyclopropyl]-13,13-difluoro-24,27-dioxo-2,17,23-trioxa-4,11,25,28-tetraazapentacyclo[26.2.1.0³,¹².0⁵,¹⁰.0¹⁸,²²]hentriaconta-3,5,7,9,11,14-hexaene-29-carboxamide |
| Product Type | API |
| Quantity | 50 mg |
| Shipping Condition | Room Temperature |