Learn More
Glecaprevir, MedChemExpress
Description
Glecaprevir is a novel HCV NS3/4A protease inhibitor, with IC50 values ranging from 3.5 to 11.3 nM. Glecaprevir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 4.09 μM.
Specifications
Specifications
| Chemical Name or Material | Glecaprevir |
| CAS | 1365970-03-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C38H46F4N6O9S |
| Quantity | 50 mg |
| Synonym | ABT-493 |
| Solubility Information | DMSO : ≥ 83.3 mg/mL (99.30 mM) |
| SMILES | O=C([C@H]1N(C([C@@H](NC(O[C@@]2([H])[C@@]3([H])CCC2)=O)C(C)(C)C)=O)C[C@@](OC4=NC5=CC=CC=C5N=C4C(F)(/C=C/CO3)F)([H])C1)N[C@]6([C@H](C(F)F)C6)C(NS(=O)(C7(CC7)C)=O)=O |
| Molecular Weight (g/mol) | 838.87 |
| Show More |