missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLB1L2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00 EUR - 624.00 EUR
Specifications
| Antigen | GLB1L2 |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18462362
|
Novus Biologicals
NBP2-31585-25ul |
25 μL |
415.00 EUR
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18164495
|
Novus Biologicals
NBP2-31585 |
0.1 mL |
624.00 EUR
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GLB1L2 Polyclonal specifically detects GLB1L2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| GLB1L2 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q8IW92 | |
| 89944 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RGRVNYGENIDDQRKGLIGNLYLNDSPLKNFRIYSLDMKKSFFQRFGLDKWSSL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| beta-galactosidase-1-like protein 2, EC 3.2.1, EC 3.2.1.-, EC 3.2.1.23, galactosidase, beta 1-like 2, MST114, MSTP114 | |
| GLB1L2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title