missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLAST Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590649
This item is not returnable.
View return policy
Description
Immunogen sequence: IVLTSVGLPT DDITLIIAVD WFLDRLRTTT NVLGDSLGAG IVEHLSRHEL KNRDVEMGNS VIEENEMKKP YQLIAQDNET EKPIDSETKM; Positive Samples: 293T, U-251MG, HeLa, H460, C6, Rat lung; Cellular Location: Membrane, Multi-pass membrane protein.
This gene encodes a member of a member of a high affinity glutamate transporter family. Mutations in this gene are associated with episodic ataxia, Type 6. Alternative splicing results in multiple transcript variants.
Specifications
| GLAST | |
| Polyclonal | |
| Unconjugated | |
| SLC1A3 | |
| AI504299; B430115D02Rik; EA6; Eaat1; EAAT-1; Excitatory amino acid transporter 1; FLJ25094; GLAST; GLAST1; GLAST-1; Glial glutamate transporter; glial high affinity glutamate transporter; GLU-T; GluT-1; glutamate transporter; glutamate/aspartate transporter; Gmt1; High-affinity neuronal glutamate transporter; MGluT1; Slc1a3; Sodium-dependent glutamate/aspartate transporter 1; solute carrier family 1 (glial high affinity glutamate transporter), member 3; solute carrier family 1 member 3; solute carrier family 1, member 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20512, 29483, 6507 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.87 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P24942, P43003, P56564 | |
| SLC1A3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 453-542 of human EAAT1/EAAT1/SLC1A3 (NP_0041633). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction