Learn More
Hecht Karl™ Glass Vials
Click to view available options
Capacity (Metric):
2 mL
60 mL
Unit Size:
Each
Pack of 690
Specifications
Specifications
| Capacity (Metric) | 60 mL |
| Material | Glass |
| Includes | Plastic snap cap |
| Packaging | 84Pack |
| Product Type | Vial |