Learn More
Hirschmann™ Glass Trapezoidal Volumetric Flask with Polyethylene Stopper
Product Code. 11746742
Click to view available options
Capacity (Metric):
1 mL
2 mL
5 mL
10 mL
20 mL
25 mL
50 mL
Standard Taper Joint:
10/19 NS Standard Ground
12/21 NS Standard Ground
7/16 NS Standard Ground
Unit Size:
Pack of 2
Product Code. 11746742
Brand: Hirschmann™ 2960133
Only null left
Add to basket
Add to basket
Manufactured using class A, DURAN™ glass
  • Blue graduation
  • Certified conformity, with dated batch identification 
  • Trapezoidal
  • Standard ground and PE stopper
  • Calibrated for IN, DIN EN ISO 1042
Specifications
Certifications/Compliance DIN EN ISO 1042
Closure Material Polyethylene
Capacity (Metric) 1 mL
Material Borosilicate Glass, Duran Glass
Height (Metric) 60 mm
Limit of Error ±0.025 mL
Standard Taper Joint 7/16 NS Standard Ground
Tolerance ±0.025 mL
Graduated Yes
Class A