Learn More
Day Impex™ Glass Tempering Beaker
Description
- Double walled glass beaker
- Includes inlet and outlet ports for controlling temperature
- Constructed of untyped glass
Specifications
Specifications
| Diameter (Metric) Outer | 70mm |
| Capacity (Metric) | 250 mL |
| Material | Glass |
| For Use With (Application) | For Heating/Cooling Function |
| Height (Metric) | 110mm |
| Quantity | 1Each |
| Type | Beaker |