Learn More
Hirschmann™ Disposable Micro Capillary Pipettes, End-to-End
Description
- End-to-end, with minicaps
- Conformity certified
- Available either heparinized or non-heparinized
- Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifications
Specifications
| Accuracy | 0.5% |
| Certifications/Compliance | ISO 7550, CE acc. IVD 98/79 EG, conformity certified |
| Description | Disposable micro capillary pipettes, not heparinized, end-to-end; includes Pipette bulb (per 1000 pcs.) |
| Material | Borosilicate Glass |
| Quantity | 10 x 100/Pack |
| Volume (Metric) | 100 μL |
| Type | Minicap Capillary Pipette |
| Disposable | Single-use |