Learn More
Karl Hecht™ Glass Raschig Rings
Product Code. 10159740
Change view
Click to view available options
Diameter (Metric):
3 mm
10 mm
Unit Size:
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10159740 3 mm 1kg
10449601 10 mm 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10159740 Supplier Karl Hecht™ Supplier No. 41400003
Only null left
Add to basket
Add to basket
For use with distillation and chemical engineering applications.
  • Provided in packs of 1kg
  • Available in two different sizes
  • Clear glass
Specifications
Type Raschig Rings
Material Glass
Diameter (Metric) 3 mm
Quantity 1kg pack