Learn More
Hamilton™ Polilyte Lab Electrode
Click to view available options
Connector Types:
S7
Unit Size:
Each
Description
- Serial number, certificate
- Well suited for measurements in emulsions and suspensions
- Thanks to the Single Pore, clogging of the liquid junction is virtually impossible
Specifications
Specifications
| Type | Polilyte Lab |
| Connector Types | S7 |
| Diameter (Metric) | 12 mm |
| Length (Metric) | 120 mm |
| Material | Glass |