Learn More
Karl Hecht™ Petri Dish, Glass
Shallow, lidded glass dishes for culturing cells including bacteria. Karl Hecht™ Petri Dishes, Glass are available in a range of convenient sizes.
Brand: Karl Hecht™ 41042036
Additional Details : Weight : 0.20000kg
Description
- Glass construction.
- Lidded.
- Available in a variety of sizes.
Specifications
Glass | |
Petri Dish | |
2.5 cm | |
15 cm |