Learn More
Hirschmann™ minicaps™ Micro Capillary Pipettes, Disposable
Product Code. p-8011841
Click to view available options
Volume (Metric):
0.5 μL
1 μL
2 μL
3 μL
4 μL
5 μL
10 μL
20 μL
25 μL
30 μL
40 μL
50 μL
100 μL
Unit Size:
Pack of 1000
Product Code. 11871060
Brand: Hirschmann™ 9000100
Only null left
Add to basket
Add to basket
Facilitates error-free capillary filling in the laboratory
  • Not heparinized
  • End-to-end, with minicaps
  • Acc. to ISO 9000, CE acc. IVD 98/79 EC
  • Color-coded vial cap with capillary dispenser
  • Conformity certified, with batch number
Specifications
Accuracy 0.5%
Certifications/Compliance ISO 7550, CE, IVD 98/79 EC
Material Glass
Volume (Metric) 0.5 μL
Length (Metric) 30 to 32 mm
Type Minicap Capillary Pipette