Learn More
Lenz Laborglasinstrumente™ Glass Distillation Thermometer
Product Code. 10529751
Change view
Click to view available options
Temperature Range (Metric):
0°C to 150°C
0°C to 250°C
Length (Metric):
55 mm
70 mm
85 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Length (Metric) unitSize
10529751 0°C to 150°C 70 mm Each
10294051 0°C to 150°C 55 mm Each
10756761 0°C to 250°C 55 mm Each
10458982 0°C to 150°C 85 mm Each
10294621 0°C to 250°C 85 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10529751 Supplier Lenz Laborglasinstrumente™ Supplier No. 05065002
Only null left
Specifications
Bottle Fill Paraffin Oil
Length (Metric) 70 mm
Material Glass
Product Type Distillation Thermometer
Max. Temperature (Metric) 150°C
Min. Temperature (Metric) 0°C
Temperature Range (Metric) 0°C to 150°C