Learn More
Hirschmann™ Glass covers
Product Code. p-7115244
Click to view available options
Dimensions (L x W):
21 x 26 mm
24 x 40 mm
24 x 60 mm
Unit Size:
Pack of 1000
Product Code. 13240480
Brand: Hirschmann™ 8000120
Only null left
Add to basket
Add to basket
Remove stickiness and minimize dust. Hirschmann™ Glass covers are perfect for protecting and preserving microscopy slides.
  • LOT number on packaging
  • CE pursuant to IVD 98/79
  • DIN 58 884
Specifications
Thickness (Metric) #1 (0.13 to 0.16 mm)
Dimensions (L x W) 24 x 60 mm