Learn More
GKN2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24651-1-AP-20UL
 View more versions of this product
Product Code. 16803145
  • 145.00 EUR / 20µL
Description
The function of this protein remains unknown.
Specifications
Specifications
GKN2
Polyclonal
Unconjugated
GKN2
BLOT, Blottin, gastrokine 2, GDDR, GKN2, PRO813, TFIZ1, VLTI465
Rabbit
Antigen Affinity Chromatography
RUO
200504, 66284
-20°C
Liquid
Western Blot
0.6 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q86XP6, Q9CQS6
Gkn2
GKN2 Fusion Protein Ag19153
20 μL
Primary
Mouse, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions