Learn More
GKN2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24651-1-AP-20UL
This item is not returnable.
View return policy
Description
The function of this protein remains unknown.Specifications
| GKN2 | |
| Polyclonal | |
| Unconjugated | |
| GKN2 | |
| BLOT, Blottin, gastrokine 2, GDDR, GKN2, PRO813, TFIZ1, VLTI465 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 200504, 66284 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q86XP6, Q9CQS6 | |
| Gkn2 | |
| GKN2 Fusion Protein Ag19153 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |