Learn More
GJ103 sodium salt, MedChemExpress
Description
GJ103 sodium salt is an active analog of the read-through compound GJ072.
Specifications
Specifications
| Chemical Name or Material | GJ103 sodium salt |
| CAS | 1459687-96-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C16H13N4NaO3S |
| Quantity | 5 mg |
| Solubility Information | DMSO : 50 mg/mL (137.23 mM; Need ultrasonic) |
| SMILES | O=C(O[Na])CSC1=NN=C(C2=NC=CC=C2)N1C3=CC=CC(OC)=C3 |
| Molecular Weight (g/mol) | 364.35 |
| Formula Weight | 364.35 |
| Show More |