Learn More
Ginsenoside Rh3, TRC
Specifications
Specifications
| Chemical Name or Material | Ginsenoside Rh3 |
| CAS | 105558-26-7 |
| Molecular Formula | C36 H60 O7 |
| Quantity | 5 mg |
| Synonym | (3beta,12beta,20Z)-12-Hydroxydammara-20(22),24-dien-3-yl beta-D-glucopyranoside |
| InChI Formula | InChI=1S/C36H60O7/c1-20(2)10-9-11-21(3)22-12-16-36(8)28(22)23(38)18-26-34(6)15-14-27(33(4,5)25(34)13-17-35(26,36)7)43-32-31(41)30(40)29(39)24(19-37)42-32/h10-11,22-32,37-41H,9,12-19H2,1-8H3/b21-11-/t22-,23-,24-,25+,26-,27+,28+,29-,30+,31-,32+,34+,35-,36-/m1/s1 |
| SMILES | CC(=CC\C=C(\C)/[C@H]1CC[C@]2(C)[C@@H]1[C@H](O)C[C@@H]3[C@@]4(C)CC[C@H](O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)C(C)(C)[C@@H]4CC[C@@]23C)C |
| IUPAC Name | (2R,3R,4S,5S,6R)-2-[[(3S,5R,8R,9R,10R,12R,13R,14R,17S)-17-[(1Z)-1,5-dimethylhexa-1,4-dienyl]-12-hydroxy-4,4,8,10,14-pentamethyl-2,3,5,6,7,9,11,12,13,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl]oxy]-6-(hydroxymethyl)tetrahydropyran-3,4,5-triol |
| Molecular Weight (g/mol) | 604.86 |
| Formula Weight | 604.4339 |