missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilteritinib hemifumarate, MedChemExpress
Description
Gilteritinib (ASP2215) hemifumarate is a potent and ATP-competitive FLT3/AXL inhibitor with IC50 of 0.29 nM/0.73 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | Gilteritinib hemifumarate |
| CAS | 1254053-84-3 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C29H44N8O3 0.5C4H4O4 |
| Quantity | 100 mg |
| Synonym | ASP2215 hemifumarate |
| Solubility Information | H2O : 2 mg/mL (3.27 mM; Need ultrasonic) ∣Ethanol : 2 mg/mL (3.27 mM; ultrasonic and warming and adjust pH to 3 with 1M HCl and heat to 60°C) ∣DMSO : 1.74 mg/mL (2.85 mM; Need ultrasonic and warming) |
| SMILES | OC(/C=C/C(O)=O)=O.NC(C1=NC(CC)=C(NC2CCOCC2)N=C1NC3=CC(OC)=C(N4CCC(N5CCN(C)CC5)CC4)C=C3)=O.[0.5] |
| Molecular Weight (g/mol) | 610.75 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?