Learn More
Macherey-Nagel™ Gilson ASPEC Chromabond™ SiOH Polypropylene SPE Column
For automated systems or vacuum manifolds
277.00 EUR - 492.00 EUR
Product Code | Brand | Column Capacity (Metric) | Bed Weight | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Column Capacity (Metric) | Bed Weight | Price | Quantity & Availability | ||||
12897911
|
Macherey-Nagel™
730071ASP |
1 mL | 100 mg |
277.00 EUR
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
12874451
|
Macherey-Nagel™
730075ASP |
6 mL | 1 g |
492.00 EUR
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
12801071
|
Macherey-Nagel™
730073ASP |
3 mL | 500 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Automated systems or CHROMABOND™ vacuum manifolds
- Polypropylene column, polyethylene filter element
- High loading capacity and separation efficiency
- Constant back pressure, airtight sealing
- Standard Luer port For automated solid phase extraction, parallel flash purification/synthesis, high throughput screening/purification.
Specifications
SPE | |
Solid Phase Extraction Column | |
Cap |