Learn More
Macherey-Nagel™ Gilson ASPEC Chromabond™ SiOH Polypropylene SPE Column
Shop All Macherey-Nagel ProductsDescription
- Automated systems or CHROMABOND™ vacuum manifolds
- Polypropylene column, polyethylene filter element
- High loading capacity and separation efficiency
- Constant back pressure, airtight sealing
- Standard Luer port For automated solid phase extraction, parallel flash purification/synthesis, high throughput screening/purification.
Specifications
Specifications
| Column Capacity (Metric) | 1 mL |
| Column Type | SPE |
| Bed Weight | 100 mg |
| Type | Solid Phase Extraction Column |
| Includes | Cap |