Learn More
Gilmont™ Gilmont™ Falling-ball Viscometer
Product Code. p-4532117 Shop All Cole Parmer Products
Change view
Click to view available options
Size:
3
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
13075706 3 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13075706 Supplier Gilmont™ Supplier No. 0870210
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Gilmont Falling-ball Viscometers offer a simple, accurate, low cost, absolute method for measuring viscosity of clear fluids up to 200cp.
Gilmont™ Falling-ball Viscometers offer a simple, accurate, low cost, absolute method for measuring viscosity of clear fluids up to 200cp. Easier to use than capillary type, and less expensive than rotating/sensor electronic-based devices.
  • Unique ball-release mechanism
  • Wetted parts of glass, PTFE and Viton™ seals (stainless steel and tantalum floats optional)
  • Three range options per tube allow for better accuracy in each range
  • Absolute reading, no electronics
  • Small sample size: approx. 7mL required
  • Easily used in constant temperature bath
  • Closed system ideal for highly evaporative and hydroscopically deteriorative solutions
Recommended for:
Used in lab, R and D, process, and plant quality control
Specifications
Product Type Falling Ball Viscometer
Viscosity Range Up to 200 cp
Size 3