missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Transferrin, Human (Custom Product)

Product Code. 11558926
Click to view available options
Quantity:
20 x 100 mL
Unit Size:
10mL
This item is not returnable. View return policy

Product Code. 11558926

Brand: Gibco 0030124SA

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

With transferrin at 4mg/mL

Cell Analysis, Cell Cycle, Cell Structure, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Lysosomes, Vacuoles

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 20 x 100 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.