Learn More
Fisherbrand™ PTFE Giant Stir Bar
Product Code. 10074400 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
57 mm
80 mm
108 mm
110 mm
136 mm
159 mm
Diameter (Metric):
14 mm
24 mm
27 mm
36 mm
Shape:
Cylinder
Cylinder, Pivot ring
Triangle
Unit Size:
Each
Pack of 5
Product Code. 10074400
Only null left
Add to basket
Add to basket
Designed for use in large containers and vials. These giant PTFE-coated magnetic stir bares are autoclavable.
  • Stable, general-purpose magnetic stir bar
  • Designed for use in large containers and vials
  • Autoclavable
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core; Cylindrical with Pivot Ring
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Diameter (Metric) 24 mm
Length (Metric) 159 mm
Quantity 1/Ea.
Shape Cylinder, Pivot ring