Learn More
Brady™ GHS Symbol - Compressed Gas, Individual Markers on a Roll
Pre-printed labels to identify your chemical product containers.
Brand: Brady™ 834185
Additional Details : Weight : 1.00000kg
Description
- High-quality labels that can withstand the most demanding environments
- Highly resistant to bad weather, abrasion and chemical products
Specifications
Gloss | |
250/Pk. | |
21 mm | |
21 mm | |
Diamond |
Polyester | |
Permanent Acrylic | |
0.099 mm | |
-40°C to 110°C | |
Black on White |