Learn More
Invitrogen™ Ghrelin Monoclonal Antibody (45)

Description
Specificity: ABS 050-45-02 binds human ghrelin and des-acyl ghrelin, and showed ~5% cross-Specificity with rat ghrelin in a sandwich ELISA using ABS 052-121 as biotinylated detection antibody. Reactivity: ABS 050-45-02 binds human ghrelin in solution. ABS 050-45-02 can be used as a capture antibody in a sandwich ELISA using biotinylated detection antibody ABS 052-121-02. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
Specifications
Specifications
| Antigen | Ghrelin |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 45 |
| Concentration | 1.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | GHRL |
| Gene Accession No. | Q9UBU3 |
| Gene Alias | 2210006E23Rik; AMCBX1; Appetite-regulating hormone; appetite-regulating hormone; uncharacterized protein LOC58991; exon 2- and 3-deleted preproghrelin; exon 2-deleted preproghrelin; FIP3; FIP-3; Fip3p; Ghr; Ghrelin; ghrelin 27; ghrelin 28; ghrelin and obestatin prepropeptide; ghrelin precursor; ghrelin, growth hormone secretagogue receptor ligand; ghrelin/obestatin; ghrelin/obestatin preprohormone; ghrelin/obestatin prepropeptide; Ghrelin-27; Ghrelin-28; ghrelin-like protein; GHRL; Growth hormone secretagogue; growth hormone-releasing peptide; IKK-gamma; In2c-preproghrelin; IP; IP1; IP2; IPD2; m46; Motilin-related peptide; MTLRP; MTLRPAP; NEMO; Obestatin; Obestatin-13; Obestatin-23; OTTHUMP00000207794; OTTHUMP00000207795; OTTHUMP00000207796; OTTHUMP00000207798; prepro-appetite regulatory hormone; preproghrelin; proghrelin; Protein M46; UNQ524/PRO1066 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.