missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Antibody, Texas Red, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB120-6660
This item is not returnable.
View return policy
Description
GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay, Knockout Validated, Knockdown Validated.
Specifications
| GFP | |
| Polyclonal | |
| Western Blot >1:10000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:500-1:2500, Fluorophore-linked immunosorbent assay >1:20000, Knockout Validated, KnockDown Validated | |
| Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Jellyfish | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoassay, KnockDown | |
| Texas Red | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide | |
| GFP | |
| The immunogen is a Green Fluorescent Protein (GFP) fusion protein corresponding to the full length amino acid sequence (246aa) derived from the jellyfish Aequorea victoria. (Uniprot: P42212) | |
| 0.2 mg | |
| Epitope Tags | |
| No reaction was observed against Human, Mouse and Rat Serum Proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction