missing translation for 'onlineSavingsMsg'
Learn More

Gestonorone Capronate, TRC

Product Code. 30132543
Change view
Click to view available options
Quantity:
100 mg
250 mg
2.5 g
Unit Size:
100mg
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132543 2.5 g 2.5g
30010692 100 mg 100mg
30012259 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132543 Supplier Toronto Research Chemicals Supplier No. TRCG3682852.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1253-28-7
Molecular Formula C26H38O4
Molecular Weight (g/mol) 414.58
Synonym 17-[(1-Oxohexyl)oxy]-19-norpregn-4-ene-3,20-dione, Hexanoic Acid Ester with 17-hydroxy-19-norpregn-4-ene-3,20-dione, 17-Hydroxy-19-norpregn-4-ene-3,20-dione Hexanoate, 17α-Hydroxy-19-norprogesterone Caproate, 17β-Acetyl-17-hydroxyestr-4-ene-3-one Hexanoate, 19-Nor-17α-caproyloxy-4-pregnene-3,20-dione, 19-Norpregn-4-ene-3,20-dione 17-Caproate, Depostat, Gestonorone Capronate, Gestronol Caproate, Gestronol Hexanoate, NSC 84054, Primostat, SH 582, SH 80582, ZK 5623
IUPAC Name [(8R,9S,10R,13S,14S,17R)-17-acetyl-13-methyl-3-oxo-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-yl] hexanoate
SMILES CCCCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C)C(=O)C

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C26H38O4/c1-4-5-6-7-24(29)30-26(17(2)27)15-13-23-22-10-8-18-16-19(28)9-11-20(18)21(22)12-14-25(23,26)3/h16,20-23H,4-15H2,1-3H3/t20-,21+,22+,23-,25-,26-/m0/s1
Formula Weight 414.277
Chemical Name or Material Gestonorone Capronate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.