Learn More
Gentamicin sulfate, MedChemExpress
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
Description
Gentamicin sulfate, an orally active aminoglycoside antibiotic, inhibits the growth of both gram-positive and gram-negative bacteria and to inhibit several strains of mycoplasma in tissue culture. Gentamicin sulfate inhibits DNase I with an IC50 of 0.57 mM.
Specifications
Specifications
| Chemical Name or Material | Gentamicin sulfate |
| CAS | 1405-41-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C(19-21)H(39-43)N5O7.H2SO4 |
| Quantity | 500 mg |
| Solubility Information | H2O : ≥ 30 mg/mL |
| SMILES | O=S(O)(O)=O.O[C@]1(C)C(NC)[C@@H](O)[C@@H](O[C@H]2C(N)C[C@H](N)[C@@H](O[C@@H]3[C@H](N)CC[C@@H]([R])O3)[C@@H]2O)OC1.NC[*].C[C@H]([*])N.C[C@H]([*])NC.[R=].[or].[or] |
| Grade | Research |
| Physical Form | Powder |
| Show More |