missing translation for 'onlineSavingsMsg'
Learn More

Gentamicin C1a Pentaacetate Salt, TRC

Product Code. 17970495
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17970495 10 mg 10mg
17900665 1 mg 1mg
17912735 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17970495 Supplier Toronto Research Chemicals Supplier No. TRCG36059010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C29H59N5O17
Molecular Weight (g/mol) 749.8
Synonym O-3-Deoxy-4-C-methyl-3-(methylamino)-β-L-arabinopyranosyl-(1-6)-O-[2,6-diamino-2,3,4,6-tetradeoxy-α-D-erythro-hexopyranosyl-(1-4)]-2-deoxy-D-streptamine Pentaacetate Salt, Gentamicin D Acetate, Gentamycin C1a Acetate, Gentamicin C3 Acetate;
SMILES NCC1O[C@H](O[C@H]2[C@@H](O)[C@H](O[C@@H]3[C@H](O)[C@@H](NC)[C@](O)(C)CO3)[C@H](N)C[C@H]2N)[C@H](N)CC1

Specifications

Analyte or Component Names Gentamicin C1a Pentaacetate Salt
Shipping Condition Room Temperature
Solution Type Neat
Industry Type Forensics
Quantity 10 mg
Percent Purity >95%
Chemical Name or Material Gentamicin C1a Pentaacetate Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.