Learn More
VITLAB™ Genius™ Dispenser
Product Code. 10622213
Click to view available options
Volume (Metric):
2.5 to 25 mL
Unit Size:
Each
Product Code. 10622213
Brand: VITLAB™ 1605506
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Accuracy ≤±0.5%
Product Type Bottletop Dispenser
Certifications/Compliance DIN EN 285, DE-M marked
Volume (Metric) 2.5 to 25 mL