missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitlab™ Genius Bottle Top Dispensers
Click to view available options
Volume (Metric):
5 to 50 mL
Unit Size:
Each
Description
- Materials that come into in contact with media (PTFE, PFA, FEP, borosilicate glass and platinum-iridium) are resistant to most acids, bases, and solvents
- Reagent loss while ventilating is avoided with the innovative recirculation valve
- Simple-to-use calibration function helps meet all the requirements for test equipment monitoring without downtime
- Sealing lip acts like a windscreen wiper to prevent crystal build-up on the cylinder wall from readily crystallisable media
- Glass cylinder is coated with a plastic material that reduces the risk of splashes should breakage occur
- Telescopic filling tube can be adjusted smoothly to different bottle heights
- Autoclavable at 121 °C (2 bar) according to DIN EN 285
- DE-M marked
- Included in delivery:
- Dispenser with 3 threaded adapters made from PP
- Nominal volumes of 2.5 – 10 ml (screw coupling GL 32) with adapters GL 28, S 40 and GL 45
- Nominal volumes of 25 – 100 ml (screw coupling GL 45) with adapters GL 32, GL 38 and S 40
- Telescopic filling tube (200 – 350 mm), mounting tool, instruction manual, and quality certificate stating all test values.
Specifications
Specifications
| Accuracy | ±0.5% |
| Product Type | Distributor |
| Autoclavable | Autoclavable |
| Certifications/Compliance | DIN EN ISO 8655-3 |
| Volume (Metric) | 5 to 50 mL |
| Increments | 1 mL |
| Warranty | 15 Months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction