missing translation for 'onlineSavingsMsg'
Learn More

Genipin 1-β-D-gentiobioside, MedChemExpress

Product Code. p-200099881
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16459258

Brand: Medchem Express HYN2094

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Genipin 1-β-D-gentiobioside (Genipin 1-gentiobioside) is one of the most abundant and bioactive iridoid glycosides in Gardenia jasminoides Ellis, which possesses hepatoprotective, anti-inflammatory, antioxidant, and antithrombotic activities.

Genipin 1-β-D-gentiobioside (Genipin 1-gentiobioside) is one of the most abundant and bioactive iridoid glycosides in Gardenia jasminoides Ellis, which possesses hepatoprotective, anti-inflammatory, antioxidant, and antithrombotic activities.

Specifications

Chemical Name or Material Genipin 1-β-D-gentiobioside
CAS 29307-60-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H34O15
Quantity 10 mg
Synonym Genipin 1-gentiobioside Genipin 1-β-gentiobioside Genipin gentiobioside
Solubility Information Ethanol : 125 mg/mL (227.06 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (181.65 mM; Need ultrasonic)
SMILES O[C@H]([C@@H](O)[C@@H]1O)[C@](O[C@@H]1CO[C@@H]([C@@H]([C@@H](O)[C@@H]2O)O)O[C@@H]2CO)([H])O[C@H]3[C@@](C(CO)=CC4)([H])[C@@]4([H])C(C(OC)=O)=CO3
Molecular Weight (g/mol) 550.51
Formula Weight 550.51
Percent Purity 98.65%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.