Learn More
Genevac™ Fixed Angle Rotor
Product Code. 10098652
Click to view available options
Capacity:
50 mL
Unit Size:
Each
Product Code. 10098652
Brand: Genevac QRC50CCT036
Specifications
Capacity 50 mL
Max. Tube Diameter 28 mm
For Use With (Equipment) 50 mL Conical Centrifuge Tubes
Angle Fixed